Learn More
Rat anti-Mouse IgG (H+L) Secondary Antibody, Invitrogen™
Rat Monoclonal Secondary Antibody
Supplier:  Thermo Scientific SA135633
Catalog No. 01-672-157
Description
Description
Thermo Scientific Anti-Mouse secondary antibodies are affinity-purified antibodies with well-characterized specificity for mouse immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Mouse IgG (H+L)
Monoclonal
1 mg/mL
Rat
Purified
RUO
Mouse
Liquid
ELISA, Immunohistochemistry (Frozen)
LO-MG-7
Unconjugated
Immunoglobulins.
1 mg
Secondary
4° C
IgM κ
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.