Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Rat anti-Mouse IgM Heavy Chain Secondary Antibody (eB121-15F9), PE, Novus Biologicals™
Rat Monoclonal Antibody
$289.00
Specifications
Antigen | IgM |
---|---|
Clone | eB121-15F9 |
Concentration | 0.2 mg/mL |
Dilution | Flow Cytometry 1:10-1:1000 |
Applications | Flow Cytometry |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP14374901
|
Novus Biologicals
NBP1437490.1MG |
0.1 mg |
Each of 1 for $289.00
|
|
|||||
Description
IgM Heavy Chain Monoclonal specifically detects IgM Heavy Chain in Mouse samples. It is validated for Flow Cytometry.Specifications
IgM | |
0.2 mg/mL | |
Flow Cytometry | |
PE | |
RUO | |
Aqueous buffer, may contain carrier protein/stabilizer (proprietary) with 0.09% Sodium Azide | |
3507 | |
The immunogen for this antibody was PE IgM. | |
Secondary | |
Store at 4C. Do not freeze. |
eB121-15F9 | |
Flow Cytometry 1:10-1:1000 | |
Monoclonal | |
Rat | |
Mouse | |
AGM1, DKFZp686I15196, DKFZp686I15212, FLJ00385, immunoglobulin heavy constant mu, MGC104996, MGC52291, MU, VH | |
IGHM | |
IgG2a κ | |
Affinity Purified |
For Research Use Only
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title