Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ly-6G Rat anti-Mouse, Brilliant Violet 510, Clone: 1A8, BD Optibuild™

Rat Monoclonal Antibody

Manufacturer:  BD Biosciences 740157

Catalog No. BDB740157


Add to cart

Description

Description

The 1A8 monoclonal antibody specifically binds to Ly-6G, a 21-25-kDa glycosyolphosphatidylinositol-anchored protein. In the bone marrow, Ly-6G is expressed on the majority of the largest cells, which are predominantly granulocytes, and not on lymphoid or erythroid cells. In the periphery, it is expressed on granulocytes. The RB6-8C5 monoclonal antibody recognizes both Ly-6G and Ly-6C and blocks the binding of mAb 1A8 to Ly-6G.

The antibody was conjugated to BD Horizon™ BV510 which is part of the BD Horizon Brilliant™ Violet family of dyes. With an Ex Max of 405-nm and Em Max at 510-nm, BD Horizon BV510 can be excited by the violet laser and detected in the BD Horizon V500 (525/50-nm) filter set. BD Horizon BV510 conjugates are useful for the detection of dim markers off the violet laser.

Specifications

Specifications

Ly-6G
1A8
Brilliant Violet 510
Aqueous buffered solution containing ≤0.09% sodium azide.
Rat
IgG2a, κ
50μg
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Monoclonal
Flow Cytometry
0.2mg/mL
Ly6g; Lymphocyte antigen 6G; Lymphocyte antigen 6 complex, locus G; Gr1
Affinity Purified
Ly-6G-transfected EL4J cell line
The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography.
RUO
Primary
Murine
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only