Learn More
Siglec-F Rat anti-Mouse, Brilliant Violet 510, Clone: E50-2440, BD Optibuild™
Rat Monoclonal Antibody
Manufacturer:  BD Biosciences 740158
Catalog No. BDB740158
Add to cart
Description
Description
The E50-2440 monoclonal antibody specifically recognizes Siglec-F. Siglecs are the U>s/U> ialic acid-binding immunoglobulin superfamily lectins defined in the human, each of which has a distinctive expression pattern in the hematopoietic system and at least some of which are known to mediate cell-cell interactions. Orthologous proteins of human Siglec-1 (Sialoadhesin or CD169), Siglec-2 (CD22), and Siglec-4 (myelin-associated glycoprotein) have been characterized in the mouse. Human Siglec-3 (CD33) and Siglecs-5 through -10 are encoded by a cluster of closely related genes, and each has two cytoplasmic ITIM (Immunoreceptor Tyrosine-based Inhibitory Motifs). Similarly, mouse Siglec-F is encoded by a gene in a syntenic cluster in the mouse, and the protein has sialic acid-binding activity and an intracytoplasmic ITIM. Its expression pattern differs from those of the human Siglec-3-related proteins in that it is found on immature cells of the myelomonocytic lineage, with reduced expression on mature neutrophils and monocytes, and not on lymphoid cells. It has been proposed that mAb E50-2440 may be used for identification of immature myelomonocytic cells in the mouse.
Specifications
Specifications
Siglec-F
E50-2440
Brilliant Violet 510
Aqueous buffered solution containing ≤0.09% sodium azide.
Rat
IgG2a, κ, also known as Louvain, LOU/C, LOU/M IgG2a, κ
50μg
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Monoclonal
Flow Cytometry
0.2mg/mL
Siglecf; Siglec5; sialic acid binding Ig-like lectin 5; SIGL5; Siglec-5
Affinity Purified
Mouse Siglec-F and human IgG Fc recombinant fusion protein
The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography.
RUO
Primary
Murine