Learn More
Rat anti-PD-L2/B7-DC/PDCD1LG2, Clone: 122, Novus Biologicals™
Manufacturer:  Novus Biologicals NBP1431160.05MG
 View more versions of this product
Catalog No. NBP14311605
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications
Specifications
PD-L2/B7-DC/PDCD1LG2
122
Unconjugated
Aqueous, may contain carrier protein/stabilizer. with 0.09% Sodium Azide
B7-DC, bA574F11.2, Btdc, Butyrophilin B7-DC, CD273 antigen, CD273PD-1 ligand 2, MGC142240, PD-1-ligand 2, PDCD1L2MGC142238, PDL2B7DC, PD-L2PDCD1 ligand 2, programmed cell death 1 ligand 2, Programmed death ligand 2
Rat
IgG2a Kappa
0.05 mg
Store at 4C. Do not freeze.
Monoclonal
Mouse
Flow Cytometry
0.5 mg/mL
Flow Cytometry 1:10-1:1000
Affinity Purified
PDCD1LG2
The immunogen for this antibody was B7DC.
Protein A or G purified
RUO
Primary
80380