Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PDGF R beta Antibody (APB5) - BSA Free, Novus Biologicals™
Rat Monoclonal Antibody has been used in 3 publications
Supplier: Novus Biologicals NBP1433490.1MG
Description
PDGF R beta Monoclonal specifically detects PDGF R beta in Mouse samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Frozen, Protein Array.Specifications
PDGF R beta | |
Monoclonal | |
0.5 mg/mL | |
Flow Cytometry < 1.0 ug/10^6 cells, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500 | |
P05622 | |
PDGFRB | |
The immunogen for this antibody was PDGF receptor beta. | |
0.1 mg | |
Protein Kinase | |
5159 | |
Store at 4C. Do not freeze. |
Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Frozen) | |
APB5 | |
Unconjugated | |
Aqueous, may contain carrier protein/stabilizer. with 0.09% Sodium Azide | |
beta-type platelet-derived growth factor receptor, CD140 antigen-like family member B, CD140b, CD140b antigen, EC 2.7.10, EC 2.7.10.1, JTK12, PDGFR, PDGFR1, PDGF-R-beta, platelet-derived growth factor receptor, beta polypeptide | |
Rat | |
Affinity Purified | |
RUO | |
Primary | |
Mouse | |
IgG2a κ |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?
Provide Content Correction