Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ Rat anti-Septin-9, Clone: 1A6C2,
Rat Monoclonal Antibody
Supplier: MilliporeSigma™ MABT321
Description
Specifically detects Septin-9 clone: 1A6C2 in Human, Mouse samples, and it is validated for Immunocytochemistry, Western Blotting
Septin-9 (also known as eighth septin, Eseptin, and MLL septin-like fusion) is encoded by the Sept9 gene in rat and mouse (SEPT9 in human). Septins are GTP-binding proteins capable of forming higher-order oligomers and filamentous polymers that contact with the cell membrane and cytoskeleton. Septins are involved in various cellular processes, including cytokinesis, karyokinesis, exocytosis, and cell shape maintenance. The expression of SEPT2, SEPT6, SEPT7 and SEPT9 in heart tissue is reported to correlate well with cardiomyocyte cytokinesis activity during heart development, is highly expressed in embryonic heart, downregulated around birth, and nondetectable in the adult heart. (PMID 16739252 & 24837489). Clone 1A6C2 was generated using an immunogen sequence present only in rat spetin-9 isoform 1 (SLP-a; Q9QZR6), but missing in rat isoform 2-4 (SLP-b, E-septin long & short forms). The immunogen sequence is 100% conserved in murine species (A2A6U3) and differs by one amino acid when compared to the corresponding human sequence in isoform 2, and missing in human isoforms 1,3,4 (Q9UHD8).
Specifications
Septin-9 | |
Monoclonal | |
Purified rat monoclonal IgG2aκ in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide. | |
41891 | |
KLH-conjugated linear peptide corresponding to the N-terminus of rat Septin-9. | |
100 μg | |
Cell Structure | |
NP_001106969 | |
Stable for one year at 2°C to 8°C from date of receipt. | |
IgG2a κ |
Immunocytochemistry, Western Blot | |
1A6C2 | |
Q9QZR6 | |
Rat | |
Protein G Purified | |
RUO | |
Primary | |
Human, Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction