Learn More
SLA (haploid sperm cell-specific), Rat, Clone: TRA54, Abnova™
Rat monoclonal antibody raised against SLA (haploid sperm cell-specific antigen).
Manufacturer:  Abnova Corporation MAB7952
Catalog No. 89-136-409
Add to cart
Description
Description
Specifications
Specifications
SLA (haploid sperm cell-specific)
TRA54
Rat monoclonal antibody raised against SLA (haploid sperm cell-specific antigen).
In PBS (50% glycerol)
Rat
IgM
RUO
Primary
Murine
Immunohistochemistry, Western Blot
Unconjugated
Immunohistochemistry Western Blot (∼1 ug/mL) The optimal working dilution should be determined by the end user.
Liquid
The hybridoma producing TRA54 was obtained from the immunaization of a rat with cell lysates of adult mouse testis.
100 ug
Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Monoclonal