Learn More
Rat anti-Thrombomodulin/BDCA-3, Clone: 4D8.2C6, Novus Biologicals™
Rat Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP236496
 View more versions of this product
Catalog No. NBP236496
Add to cart
Description
Description
Thrombomodulin/BDCA-3 Monoclonal antibody specifically detects Thrombomodulin/BDCA-3 in Mouse samples. It is validated for Western Blot, Flow Cytometry.
Specifications
Specifications
Thrombomodulin/BDCA-3
4D8.2C6
Unconjugated
PBS with 0.05% Sodium Azide
BDCA3, CD141, CD141 antigen, fetomodulin, thrombomodulin, TM
Rat
IgG1
0.1 mg
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Monoclonal
Mouse
Flow Cytometry, Western Blot
1.0 mg/mL
Western Blot 2 ug/ml, Flow Cytometry 2 ug 1 million cells
Purified
THBD
Two synthetic peptides made to internal portions of the mouse Thrombomodulin protein (between residues 25-150 and 400-550) [UniProt P07204]
Protein G purified
RUO
Primary
7056