Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thrombomodulin/BDCA-3 Antibody (4D8.2C6) - BSA Free, Novus Biologicals™
SDP

Rat Monoclonal Antibody

$204.00 - $409.50

Specifications

Antigen Thrombomodulin/BDCA-3
Clone 4D8.2C6
Concentration 1.0 mg/mL
Dilution Western Blot 2 ug/ml, Flow Cytometry 2 ug 1 million cells
Applications Western Blot, Flow Cytometry
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP236496SS
SDP
View Documents
Novus Biologicals
NBP236496SS
0.025 mg
Each for $204.00
Add to Cart
 
NBP236496
SDP
View Documents
Novus Biologicals
NBP236496
0.1 mg
Each for $409.50
Add to Cart
 
Description

Description

Thrombomodulin/BDCA-3 Monoclonal specifically detects Thrombomodulin/BDCA-3 in Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry-Paraffin.
Specifications

Specifications

Thrombomodulin/BDCA-3
1.0 mg/mL
Western Blot, Flow Cytometry
Unconjugated
Rat
Cardiovascular Biology
PBS with 0.05% Sodium Azide
7056
Two synthetic peptides made to internal portions of the mouse Thrombomodulin protein (between residues 25-150 and 400-550) [UniProt P07204]
Primary
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
4D8.2C6
Western Blot 2 ug/ml, Flow Cytometry 2 ug 1 million cells
Monoclonal
Purified
RUO
Mouse
BDCA3, CD141, CD141 antigen, fetomodulin, thrombomodulin, TM
THBD
IgG1
Protein G purified
SDS
Documents

Documents

Product Certifications

For Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit