Learn More
Rat anti-TLR9, Clone: eB72-1665, Novus Biologicals™
Rat Monoclonal Antibody
Manufacturer:  Novus Biologicals NBP1431400.025MG
 View more versions of this product
Catalog No. NBP14314003
Add to cart
Description
Description
TLR9 Monoclonal antibody specifically detects TLR9 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin.
Specifications
Specifications
TLR9
eB72-1665
Unconjugated
Aqueous, may contain carrier protein/stabilizer. with 0.09% Sodium Azide
Affinity Purified
TLR9
The immunogen for this antibody was Tolllike receptor 9.
Protein A or G purified
RUO
Store at 4C. Do not freeze.
Monoclonal
Human
Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
0.5 mg/mL
Western Blot 1:100-1:2000, Flow Cytometry 1 ug/10^6 cells in 100 ul, Immunohistochemistry 1:10-1:500, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500
Q9NR96
CD289, CD289 antigen, toll-like receptor 9
Rat
IgG2a Kappa
0.025 mg
Cytokine Research, Immunology, Innate Immunity, Toll Like Receptors
Primary
54106