Learn More
Rat anti-TRAIL/TNFSF10, Clone: N2B2, Novus Biologicals™
Manufacturer:  Novus Biologicals NBP1432720.5MG
 View more versions of this product
Catalog No. NBP1432725
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications
Specifications
TRAIL/TNFSF10
N2B2
Unconjugated
Aqueous, may contain carrier protein/stabilizer. with 0.09% Sodium Azide
Apo-2 ligand, APO2L, Apo-2Ltumor necrosis factor (ligand) family, member 10, APO2Ltumor necrosis factor apoptosis-inducing ligand splice variant delta, CD253, CD253 antigen, Protein TRAIL, TL2, TNF-related apoptosis-inducing ligand, TRAILTNF-related apoptosis inducing ligand TRAIL, tumor necrosis factor (ligand) superfamily, member 10, tumor necrosis factor ligand superfamily member 10
Rat
IgG2a Kappa
0.5 mg
Apoptosis, Cancer, Cell Biology, Signal Transduction, Tumor Suppressors
Primary
8743
Flow Cytometry, Immunoprecipitation, Functional Assay
0.2 mg/mL
Flow Cytometry 1:10-1:1000, Immunoprecipitation 1:10-1:500, Functional
Affinity Purified
TNFSF10
The immunogen for this antibody was Trail.
Protein A or G purified
RUO
Store at 4C. Do not freeze.
Monoclonal
Mouse