Learn More
Rat anti-VEGF R3/Flt-4, Clone: AFL4, Biotin, Novus Biologicals™
Rat Monoclonal Antibody
Manufacturer:  Novus Biologicals NBP1435020.05MG
 View more versions of this product
Catalog No. NBP14350205
Add to cart
Description
Description
VEGF R3/Flt-4 Monoclonal antibody specifically detects VEGF R3/Flt-4 in Mouse, Reptile samples. It is validated for Flow Cytometry.
Specifications
Specifications
VEGF R3/Flt-4
AFL4
Flow Cytometry 1:10-1:1000
Affinity Purified
FLT4
The immunogen for this antibody was VEGFR3.
Protein A or G purified
RUO
Store at 4C. Do not freeze.
Monoclonal
Mouse
Flow Cytometry
Biotin
Aqueous buffer, may contain carrier protein/stabilizer (proprietary) with 0.09% Sodium Azide
EC 2.7.10, EC 2.7.10.1, FLT-4, fms-related tyrosine kinase 4, LMPH1A, PCLFLT41, soluble VEGFR3 variant 1, soluble VEGFR3 variant 2, soluble VEGFR3 variant 3, Tyrosine-protein kinase receptor FLT4, vascular endothelial growth factor receptor 3, VEGFR-3, VEGFR3Fms-like tyrosine kinase 4
Rat
IgG2a Kappa
0.05 mg
Angiogenesis, Cancer, Hypoxia, Protein Kinase
Primary
2324