Learn More
Novus Biologicals™ Rat beta-1 Adrenergic R/ADRB1 ELISA Kit (Colorimetric)
ELISA kits are ideal for the quantification of target antigens in tissue extracts, serum and cell culture.
Manufacturer: Novus Biologicals™ NBP266654
Description
The Rat beta-1 Adrenergic R/ADRB1 ELISA Kit (Colorimetric) measures rat beta-1 Adrenergic R/ADRB1 in serum, plasma and other biological fluids.Specifications
78.13 - 5000 pg/mL | |
This kit recognizes Rat ADR(beta)1 in samples. No significant cross-reactivity or interference between Rat ADR(beta)1 and analogues was observed. | |
153 | |
Rat | |
1 Kit | |
Storage is content dependent. |
46.88 pg/mL | |
ELISA | |
ADRB1 | |
ELISA Kit (Colorimetric) | |
Biological Fluids,Plasma,Serum | |
beta-1 Adrenergic R/ADRB1 |