Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Rat bFGF ELISA Kit Non-distribution product as customer accommodation.

Rat bFGF quantitates rat bFGF in serum, plasma, supernatant.

$491.00 - $2205.00

Specifications

Regulatory Status Research Use Only
Storage Requirements 2°C to 8°C
Target FGF2
Product Type ELISA
Kit Contents Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
ERA4RBX5 Non-distribution product as customer accommodation.
View Documents
Invitrogen™
ERA4RBX5
5 x 96 Tests
Each for $2,205.00
Add to cart
 
ERA4RB Non-distribution product as customer accommodation.
View Documents
Invitrogen™
ERA4RB
96 Tests
Each for $491.00
Add to cart
 
Description

Description

Rat bFGF quantitates rat bFGF in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant rat bFGF.

FGF2 (FGFb, fibroblast growth factor basic) belongs to the fibroblast growth factor (FGF) family, and interacts with high-affinity transmembrane receptors to influence cell proliferation and tissue neovascularization. FGF2 exists as five isoforms with distinct intracellular localizations and functions. The 18 kDa isoform is predominantly cytosolic and acts through cell surface receptors, whereas the 22, 22.5, 24 and 34 kDa isoforms are nuclear and may signal independent of transmembrane receptor pathways. In humans, the gene is located on the q arm of chromosome 4. FGF2 has been implicated in diverse biological processes, such as limb and nervous system development, wound healing, and tumor growth. The mRNA for FGF2 contains multiple polyadenylation sites, and is alternatively translated from non-AUG and AUG initiation codons, resulting in five different isoforms with distinct properties. The CUG-initiated isoforms are localized in the nucleus and are responsible for the intracrine effect, whereas, the AUG-initiated form is mostly cytosolic and is responsible for the paracrine and autocrine effects of this FGF. Diseases associated with FGF2 dysfunction include Kaposi Sarcoma and corneal neovascularization.
Specifications

Specifications

Research Use Only
FGF2
Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
4 hr 45 min
Serum, Plasma, Supernatant
2°C to 8°C
ELISA
2 pg/mL
1 hr 20 min
Plasma, 50 µL; Supernatant, 50 µL; Serum, 50 µL
SDS
Product Certifications
Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit