Learn More
Novus Biologicals™ Rat CAIN ELISA Kit (Colorimetric)
ELISA kits are ideal for the quantification of target antigens in tissue extracts, serum and cell culture.
Manufacturer:  Novus Biologicals™ NBP274960
Catalog No. NBP274960
Add to cart
Description
Description
The Rat CAIN ELISA Kit (Colorimetric) measures rat CAIN in serum, plasma and other biological fluids.
Specifications
Specifications
0.16 - 10 ng/mL
ELISA
ELISA Kit (Colorimetric)
Storage is content dependent.
Rat
1 Kit
This kit recognizes Rat CABIN1 in samples. No significant cross-reactivity or interference between Rat CABIN1 and analogues was observed.
23523
0.10 ng/mL
CAIN
CABIN1
Biological Fluids, Plasma, Serum