Learn More
Abnova™ Rat Casp3 (P55213) Recombinant Protein
Used for Func, SDS-PAGE
Manufacturer:  Abnova™ P5129
Catalog No. 89-938-082
Add to cart
Description
Description
Specifications
Specifications
Escherichia coli expression system
Functional Study, Sodium Dodecyl Sulfate–Polyacrylamide Gel Electrophoresis
Lyophilized
11, 17kDa
Escherichia coli expression system
Store at -80°C on dry atmosphere. After reconstitution with PBS containing 15% glycerol 1 uL per unit, store at -80°C. Aliquot to avoid repeated freezing and thawing.
Lice/MGC93645
Casp3
Escherichia coli
None
P55213
Lyophilized
25402
Casp3 (Rat) Recombinant Protein
25 Units
RUO
Casp3
300000 units/mg
Yes