Learn More
Novus Biologicals™ Rat Cathepsin B ELISA Kit (Colorimetric)
ELISA kits are ideal for the quantification of target antigens in tissue extracts, serum and cell culture.
Supplier: Novus Biologicals™ NBP267255
Description
The Rat Cathepsin B ELISA Kit (Colorimetric) measures rat Cathepsin B in serum, plasma and other biological fluids.Specifications
0.16 - 10 ng/mL | |
This kit recognizes Rat CTSB in samples. No significant cross-reactivity or interference between Rat CTSB and analogues was observed. | |
Biological Fluids, Plasma, Serum | |
1508 | |
Rat | |
Storage of components varies. See protocol for specific instructions. |
0.10 ng/mL | |
ELISA Kit (Colorimetric) | |
ELISA | |
CTSB | |
1 Kit | |
Cathepsin B |