Learn More
Novus Biologicals™ Rat CD3 ELISA Kit (Colorimetric)
ELISA kits are ideal for the quantification of target antigens in tissue extracts, serum and cell culture.
Manufacturer:  Novus Biologicals™ NBP275138
Catalog No. NBP275138
Add to cart
Description
Description
The Rat CD3 ELISA Kit (Colorimetric) measures rat CD3 in serum, plasma and other biological fluids.
Specifications
Specifications
0.31 - 20 ng/mL
ELISA
ELISA Kit (Colorimetric)
Storage is content dependent.
Rat
1 Kit
This kit recognizes Rat CD3 in samples. No significant cross-reactivity or interference between Rat CD3 and analogues was observed.
916
0.19 ng/mL
CD3
CD3E
Biological Fluids, Plasma, Serum