Learn More
ACROBiosystems Rat CD73 Protein, His Tag (active enzyme)
Supplier:  ACROBiosystems CD3R52H520UG
Encompass_Preferred
Rat CD73, His Tag (CD3-R52H5) is expressed from human 293 cells (HEK293). It contains AA Trp 27 - Ser 551 (Accession # P21588).
Catalog No. 50-233-1707