Learn More
Novus Biologicals™ Rat CETP ELISA Kit (Colorimetric)
ELISA kits are ideal for the quantification of target antigens in tissue extracts, serum and cell culture.
Manufacturer:  Novus Biologicals™ NBP275241
Catalog No. NBP275241
Add to cart
Description
Description
The Rat CETP ELISA Kit (Colorimetric) measures rat CETP in serum, plasma and other biological fluids.
Specifications
Specifications
0.16 - 10 ng/mL
ELISA
ELISA Kit (Colorimetric)
Storage is content dependent.
Rat
1 Kit
This kit recognizes Rat CETP in samples. No significant cross-reactivity or interference between Rat CETP and analogues was observed.
1071
0.10 ng/mL
CETP
CETP
Biological Fluids, Plasma, Serum