Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Rat CINC-2 ELISA Kit Non-distribution product as customer accommodation.

Rat CINC-2 quantitates rat CINC-2 in serum, plasma, supernatant.

$491.00 - $2205.00

Specifications

Regulatory Status Research Use Only
Storage Requirements 2°C to 8°C
Target CXCL3
Product Type ELISA
Kit Contents Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
ERA7RBX5 Non-distribution product as customer accommodation.
View Documents
Invitrogen™
ERA7RBX5
5 x 96 Tests
Each for $2,205.00
Add to cart
 
ERA7RB Non-distribution product as customer accommodation.
View Documents
Invitrogen™
ERA7RB
96 Tests
Each for $491.00
Add to cart
 
Description

Description

Rat CINC-2 quantitates rat CINC-2 in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant rat CINC-2.

CXCL3 may play a role in inflammation and exert its effects on endothelial cells in an autocrine fashion.
Specifications

Specifications

Research Use Only
CXCL3
Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
4 hr 45 min
Serum, Plasma, Supernatant
2°C to 8°C
ELISA
0.05 ng/mL
1 hr 20 min
Plasma, 50 µL; Supernatant, 50 µL; Serum, 50 µL
SDS
Product Certifications
Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit