Learn More
Novus Biologicals™ Rat Clusterin/APOJ ELISA Kit (Colorimetric)
ELISA kits are ideal for the quantification of target antigens in tissue extracts, serum and cell culture.
Manufacturer:  Novus Biologicals™ NBP275388
Catalog No. NBP275388
Add to cart
Description
Description
The Rat Clusterin/APOJ ELISA Kit (Colorimetric) measures rat Clusterin/APOJ in serum, plasma and other biological fluids.
Specifications
Specifications
1.56 - 100 ng/mL
ELISA
ELISA Kit (Colorimetric)
Storage is content dependent.
Rat
1 Kit
This kit recognizes Rat CLU in samples. No significant cross-reactivity or interference between Rat CLU and analogues was observed.
1191
0.94 ng/mL
Clusterin/APOJ
CLU
Biological Fluids, Plasma, Serum