Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Gibco™ Rat CNTF Recombinant Protein, PeproTech®
E. coli Recombinant Protein
$89.50 - $3325.00
Specifications
Accession Number | P20294 |
---|---|
For Use With (Application) | ELISA, Functional Assay, Western Blot |
Formulation | protein with no preservative |
Gene ID (Entrez) | 25707 |
Molecular Weight (g/mol) | 22.7 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
45050100UG
|
Gibco™
45050100UG |
100 μg |
Each for $664.00
|
|
|||||
450-50-1MG
|
Gibco™
450501MG |
1 mg |
Each for $3,325.00
|
|
|||||
45050250UG
|
Gibco™
45050250UG |
250 μg |
Each for $1,164.00
|
|
|||||
450-50-25UG
|
Gibco™
4505025UG |
25 μg |
Each for $217.00
|
|
|||||
45050500UG
|
Gibco™
45050500UG |
500 μg |
Each for $1,994.00
|
|
|||||
450-50-50UG
|
Gibco™
4505050UG |
50 μg |
Each for $399.00
|
|
|||||
450-50-5UG
|
Gibco™
450505UG |
5 μg |
Each for $89.50
|
|
|||||
Description
The protein encoded by this gene is a polypeptide hormone whose actions appear to be restricted to the nervous system where it promotes neurotransmitter synthesis and neurite outgrowth in certain neuronal populations. The protein is a potent survival factor for neurons and oligodendrocytes and may be relevant in reducing tissue destruction during inflammatory attacks. A mutation in this gene, which results in aberrant splicing, leads to ciliary neurotrophic factor deficiency, but this phenotype is not causally related to neurologic disease. A read-through transcript variant composed of ZFP91 and CNTF sequence has been identified, but it is thought to be non-coding. Read-through transcription of ZFP91 and CNTF has also been observed in mouse.Specifications
P20294 | |
protein with no preservative | |
22.7 | |
E. coli | |
RUO | |
AI429687; Ciliary neurotrophic factor; ciliary neurotropic factor; Cntf; HCNTF; H-CNTF | |
Rat | |
Recombinant | |
Lyophilized |
ELISA, Functional Assay, Western Blot | |
25707 | |
Rat CNTF | |
-20°C | |
<1 EU/μg | |
CNTF | |
Unconjugated | |
AFAEQTPLTL HRRDLCSRSI WLARKIRSDL TALMESYVKH QGLNKNINLD SVDGVPVAST DRWSEMTEAE RLQENLQAYR TFQGMLTKLL EDQRVHFTPT EGDFHQAIHT LMLQVSAFAY QLEELMVLLE QKIPENEADG MPATVGDGGL FEKKLWGLKV LQELSQWTVR SIHDLRVISS HQMGISALES HYGAKDKQM | |
≥ 98% by SDS-PAGE gel and HPLC analyses. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title