Learn More
Novus Biologicals™ Rat Collagen II ELISA Kit (Colorimetric)
ELISA kits are ideal for the quantification of target antigens in tissue extracts, serum and cell culture.
Manufacturer:  Novus Biologicals™ NBP275854
Catalog No. NBP275854
Add to cart
Description
Description
The Rat Collagen II ELISA Kit (Colorimetric) measures rat Collagen II in serum, plasma and other biological fluids.
Specifications
Specifications
62.50 - 4000 pg/mL
ELISA
ELISA Kit (Colorimetric)
Storage is content dependent.
Rat
1 Kit
This kit recognizes Rat COL2a1 in samples. No significant cross-reactivity or interference between Rat COL2a1 and analogues was observed.
1280
37.50 pg/mL
Collagen II
COL2A1
Biological Fluids, Plasma, Serum