Learn More
Novus Biologicals™ Rat Collagen III alpha 1/COL3A1 ELISA Kit (Colorimetric)
ELISA kits are ideal for the quantification of target antigens in tissue extracts, serum and cell culture.
Manufacturer:  Novus Biologicals™ NBP275860
Catalog No. NBP275860
Add to cart
Description
Description
The Rat Collagen III alpha 1/COL3A1 ELISA Kit (Colorimetric) measures rat Collagen III alpha 1/COL3A1 in serum, plasma and other biological fluids.
Specifications
Specifications
0.63 - 40 ng/mL
ELISA
ELISA Kit (Colorimetric)
Storage is content dependent.
Rat
1 Kit
This kit recognizes Rat COL3a1 in samples. No significant cross-reactivity or interference between Rat COL3a1 and analogues was observed.
1281
0.38 ng/mL
Collagen III alpha 1/COL3A1
COL3A1
Biological Fluids, Plasma, Serum