Learn More
Novus Biologicals Rat Connexin 31/GJB3 ELISA Kit (Colorimetric)
This kit recognizes Rat CX31 in samples.
Manufacturer: Novus Biologicals NBP279815
Description
No Significant cross-reactivity or interference between Rat CX31 and analogues was observed.Specifications
0.63 to 40 ng/mL | |
2707 | |
0.38 ng/mL | |
Rat | |
1 Kit | |
Serum, plasma, and other biological fluids |
ELISA | |
ELISA Kit (Colorimetric) | |
Storage is content dependent | |
GJB3 | |
Connexin 31/GJB3 |