Learn More
Novus Biologicals™ Rat Contactin-4 ELISA Kit (Colorimetric)
ELISA kits are ideal for the quantification of target antigens in tissue extracts, serum and cell culture.
Manufacturer:  Novus Biologicals™ NBP275812
Catalog No. NBP275812
Add to cart
Description
Description
The Rat Contactin-4 ELISA Kit (Colorimetric) measures rat Contactin-4 in serum, plasma and other biological fluids.
Specifications
Specifications
0.31 - 20 ng/mL
ELISA
ELISA Kit (Colorimetric)
Storage is content dependent.
Rat
1 Kit
This kit recognizes Rat CNTN4 in samples. No significant cross-reactivity or interference between Rat CNTN4 and analogues was observed.
152330
0.19 ng/mL
Contactin-4
CNTN4
Biological Fluids, Plasma, Serum