Learn More
Abnova™ Rat Cpb1 (P19223) Partial Recombinant Protein
Used for Func, SDS-PAGE
Manufacturer:  Abnova™ P6074
Catalog No. 89-940-328
Add to cart
Description
Description
Sequence: ASGHSYTKYNNWETIEAWIQQVATDNPDLVTQSVIGTTFEGRNMYVLKIGKTRPNKPAIFIDCGFHAREWISPAFCQWFVREAVRTYNQEIHMKQLLDELDFYVLPVVNIDGYVYTWTKDRMWRKTRSTMAGSSCLGVDPNRNFNAGWCEVGASRSPCSETYCGPAPESEKETKALADFIRNNLSTIKAYLTIHSYSQMMLYPYSYDYKLPENYEELNALVKGAAKELATLHGTKYTYGPGATTIYPAAGGSDDWSYDQGIKYSFTFELRDTGFFGFLLPESQIRQTCEETMLAVKYIANYVREHLY