Learn More
Novus Biologicals Rat CREB ELISA Kit (Colorimetric)
ELISA kits are ideal for the quantification of target antigens in tissue extracts, serum and cell culture.
Manufacturer: Novus Biologicals NBP267251
Description
The Rat CREB ELISA Kit (Colorimetric) measures rat CREB in serum, plasma and other biological fluids.Specifications
ELISA Kit (Colorimetric) | |
This kit recognizes Rat CREB in samples. No significant cross-reactivity or interference between Rat CREB and analogues was observed. | |
1385 | |
CREB | |
1 Kit | |
0.19 pmol/mL |
0.31 - 20 pmol/mL | |
ELISA | |
Storage is content dependent. | |
Rat | |
Biological Fluids, Plasma, Serum | |
CREB1 |