Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Rat Cx3cl1 (O55145) Partial Recombinant Protein

Used for Func, SDS-PAGE

Manufacturer:  Abnova™ P5998

Catalog No. 89-941-019


Add to cart

Description

Description

Sequence: QHLGMTKCNITCHKMTSPIPVTLLIHYQLNQESCGKRAIILETRQHRHFCADPKEKWVQDAMKHLDHQTAALTRNG
Specifications

Specifications

Escherichia coli expression system
Functional Study, Sodium Dodecyl Sulfate–Polyacrylamide Gel Electrophoresis
Lyophilized
8.7kDa
Escherichia coli expression system
20 ug
Store at -20°C. Aliquot to avoid repeated freezing and thawing.
<0.1ng/μg (<1 EU/μg).
Cx3cl1
Activity assay
SDS-PAGE
The optimal working dilution should be determined by the end user.
Rat ;Rat
Yes
O55145
Lyophilized
89808
Cx3cl1 (Rat) Recombinant Protein
0.98
QHLGMTKCNITCHKMTSPIPVTLLIHYQLNQESCGKRAIILETRQHRHFCADPKEKWVQDAMKHLDHQTAALTRNG
RUO
Cx3c/Scyd1
Cx3cl1
Determined by its ability to chemoattract human monocytes using a concentration range of 5.0-10.0ng/ml.
Escherichia coli
None
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit