Learn More
Novus Biologicals™ Rat CXCL14/BRAK ELISA Kit (Colorimetric)
SDP
ELISA kits are ideal for the quantification of target antigens in tissue extracts, serum and cell culture.
Supplier:  Novus Biologicals™ NBP270016
Catalog No. NBP270016
Add to Cart
Description
Description
The Rat CXCL14/BRAK ELISA Kit (Colorimetric) measures rat CXCL14/BRAK in serum, plasma and other biological fluids.
Specifications
Specifications
93.75 - 6000 pg/mL
This kit recognizes Rat BRAK/CXCL14 in samples. No significant cross-reactivity or interference between Rat BRAK/CXCL14 and analogues was observed.
Biological Fluids, Plasma, Serum
9547
Rat
Storage of components varies. See protocol for specific instructions.
56.25 pg/mL
ELISA Kit (Colorimetric)
ELISA
CXCL14
1 Kit
CXCL14/BRAK
Product Suggestions
Product Suggestions