Learn More
Novus Biologicals™ Rat Cytokeratin 18 ELISA Kit (Colorimetric)
ELISA kits are ideal for the quantification of target antigens in tissue extracts, serum and cell culture.
Manufacturer:  Novus Biologicals™ NBP275285
Catalog No. NBP275285
Add to cart
Description
Description
The Rat Cytokeratin 18 ELISA Kit (Colorimetric) measures rat Cytokeratin 18 in serum, plasma and other biological fluids.
Specifications
Specifications
0.16 - 10 ng/mL
ELISA
ELISA Kit (Colorimetric)
Storage is content dependent.
Rat
1 Kit
This kit recognizes Rat CK-18/KRT18 in samples. No significant cross-reactivity or interference between Rat CK-18/KRT18 and analogues was observed.
3875
0.10 ng/mL
Cytokeratin 18
KRT18
Biological Fluids, Plasma, Serum