Learn More
Novus Biologicals™ Rat Dopamine D3R/DRD3 ELISA Kit (Colorimetric)
SDP
ELISA kits are ideal for the quantification of target antigens in tissue extracts, serum and cell culture.
Supplier:  Novus Biologicals™ NBP267940
Catalog No. NBP267940
Add to Cart
Description
Description
The Rat Dopamine D3R/DRD3 ELISA Kit (Colorimetric) measures rat Dopamine D3R/DRD3 in serum, plasma and other biological fluids.
Specifications
Specifications
0.31 - 20 ng/mL
This kit recognizes Rat DRD3 in samples. No significant cross-reactivity or interference between Rat DRD3 and analogues was observed.
Biological Fluids, Plasma, Serum
1814
Rat
GPCR
Dopamine D3 R/DRD3
0.19 ng/mL
ELISA Kit (Colorimetric)
ELISA
DRD3
1 Kit
Storage of components varies. See protocol for specific instructions.
Product Suggestions
Product Suggestions