Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Abnova™ Rat Egf (P07522, 53 amino acids) Partial Recombinant Protein
Used for Func, SDS-PAGE
Supplier: Abnova™ P5138
Specifications
P07522 | |
Lyophilized | |
6.151kDa | |
Escherichia coli expression system | |
Store at -20°C on dry atmosphere. After reconstitution with sterilized water, store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
Egf | |
The ED50 as calculated by the dose-dependant proliferation of murine BALB/c 3T3 cells is less than 0.1ng/mL, corresponding to a specific activity of ≥ 1 x 107 units/mg. | |
Recombinant | |
Escherichia coli expression system | |
≥98% by SDS-PAGE and RP-HPLC |
Functional Study, SDS-PAGE | |
25313 | |
Egf (Rat) Recombinant Protein | |
20 ug | |
RUO | |
Egf | |
E. coli | |
None | |
Lyophilized |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction