Learn More
Raybiotech IncSupplier Diversity Partner Rat FGF-BP ELISA Kit DFS Item
Manufacturer:  Raybiotech IncSupplier Diversity Partner ELRFGFBP1
For Serum, Plasma, and Cell Culture Supernatants. Sensitivity: 0.41 ng/ml. Pre-coated 96-well strip plate with additional reagents to allow for use in multiple experiments. Specificity: This ELISA antibody pair recognizes Rat FGF-BP.
Catalog No. 50-195-0241