Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rat GM-CSF (aa 18-144), Recombinant Protein, Invitrogen™ Non-distribution product as customer accommodation.

E. coli Recombinant Protein

$160.50 - $2400.00

Specifications

Recombinant Yes
Name Rat GM-CSF (aa 18-144)
For Use With (Application) Control, ELISA, Western Blot
Concentration 200 µg/mL
Conjugate Unlabeled
View More Specs

Products 4
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
PIRP87916 Non-distribution product as customer accommodation.
View Documents
Thermo Scientific
RP87916
10 µg
Each for $160.50
Add to cart
 
PIRP87917 Non-distribution product as customer accommodation.
View Documents
Thermo Scientific
RP87917
25 µg
Each for $276.50
Add to cart
 
PIRP87918 Non-distribution product as customer accommodation.
View Documents
Thermo Scientific
RP87918
100 µg
Each for $802.50
Add to cart
 
PIRP87919 Non-distribution product as customer accommodation.
View Documents
Thermo Scientific
RP87919
500 µg
Each for $2,400.00
Add to cart
 
Description

Description

GM-CSF (Granulocyte-Macrophage colony-stimulating factor) is a 14.6kDa hematopoietic growth factor that exists in glycosylated and non-glycosylated biologically active forms, and stimulates the development of granulocytes, macrophages, early megakaryocytes and eosinophil progenitor cells. The active form of the GM-CSF protein is found extracellularly as a homodimer and the GM-CSF gene has been localized to a cluster of related genes at chromosome region 5q31, which is known to be associated with interstitial deletions in the 5q-syndrome and acute myelogenous leukemia. Other genes in the cluster include those encoding interleukins 4, 5, and 13. The ability of recombinant GM-CSF to increase hematopoietic cell recovery has become a focus area in the therapeutic treatment of patients following bone marrow transplantation. Recent studies have investigated GM-CSF in inflammatory and autoimmune diseases such as arthritis, arthritis related interstitial lung disease, nephritis, and psoriasis. In the CNS, GM-CSF depletion or neutralization has been studied in experimental autoimmune encephalomyelitis (EAE) and multiple sclerosis (MS).
Specifications

Specifications

Yes
Control, ELISA, Western Blot
Unlabeled
PBS with no preservative; pH 7.2
E. coli
-20°C
P48750
CSF2
>95%
Rat GM-CSF (aa 18-144)
200 µg/mL
Liquid
colony stimulating factor 2; colony stimulating factor 2 (granulocyte-macrophage); colony-stimulating factor; CSF; CSF2; Csfgm; cytokine; GMCSF; Gm-CSf; granulocyte macrophage colony stimulating factor; granulocyte macrophage-colony stimulating factor; granulocyte-macrophage colony stimulating factor 2; granulocyte-macrophage colony-stimulating factor; MGC131935; MGC138897; MGC151255; MGC151257; MGI-IGM; M-GM-CSF; Molgramostin; put. GM-CSF; sargramostim
RUO
Rat
116630
14.6 kD
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit