Learn More
Novus Biologicals Rat GRK2 ELISA Kit (Colorimetric)
ELISA kits are ideal for the quantification of target antigens in tissue extracts, serum and cell culture.
Manufacturer: Novus Biologicals NBP266688
Description
The Rat GRK2 ELISA Kit (Colorimetric) measures rat GRK2 in serum, plasma and other biological fluids.Specifications
ELISA Kit (Colorimetric) | |
This kit recognizes Rat GRK2 in samples. No significant cross-reactivity or interference between Rat GRK2 and analogues was observed. | |
156 | |
GRK2 | |
1 Kit | |
0.10 ng/mL |
0.16 - 10 ng/mL | |
ELISA | |
Storage is content dependent. | |
Rat | |
Biological Fluids, Plasma, Serum | |
GRK2 |