Learn More
Crystal Chem IncSupplier Diversity Partner Rat Haptoglobin ELISA Kit, 96-wells DFS Item
Manufacturer:  Crystal Chem IncSupplier Diversity Partner 80677
The Rat Haptoglobin ELISA is used to quantify levels of rat haptoglobin in serum and plasma using only a 5 µL sample volume. Highlights: Kit uses sample size of only 5 µL. High sensitivity of 1.05 ng/mL. Dynamic range of 1.95 - 125 ng/mL. Highly precise with low CV's. Crystal Chem is a worldwide leading supplier of mouse and rat ELISA kits for research use. Our kits are among the highest quality found in the market, and are used by leading research universities, pharma companies, and government institutions around the world.
Catalog No. 50-194-7804