Learn More
Novus Biologicals Rat HO-1/HMOX1/HSP32 ELISA Kit (Colorimetric)
ELISA kits are ideal for the quantification of target antigens in tissue extracts, serum and cell culture.
Manufacturer: Novus Biologicals NBP262148
Description
The Rat HO-1/HMOX1/HSP32 ELISA Kit (Colorimetric) measures rat HO-1/HMOX1/HSP32 in cell lysates, plasma, serum, tissue, bronchoalveolar lavage fluid.Specifications
ELISA Kit (Colorimetric) | |
ELISA | |
Storage of components varies. See protocol for specific instructions. | |
Rat | |
Bronchoalveolar Lavage Fluid, Cell Lysates, Plasma, Serum, Tissue | |
HMOX1 |
0.195 - 12.5 ng/ml | |
3162 | |
HO-1/HMOX1/HSP32 | |
1 Kit | |
0.036 ng/ml |