Learn More
Novus Biologicals™ Rat Hsp47 ELISA Kit (Colorimetric)
Rat Hsp47 ELISA Kit (Colorimetric) from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.
Manufacturer:  Novus Biologicals™ NBP276581
Catalog No. NBP276581
Add to cart
Description
Description
The Rat Hsp47 ELISA Kit (Colorimetric) measures rat Hsp47 in serum, plasma and other biological fluids.
Specifications
Specifications
1.56 to 100 ng/mL
ELISA Kit (Colorimetric)
This kit recognizes Rat HSP47 in samples. No significant cross-reactivity or interference between Rat HSP47 and analogues was observed.
Rat
1 Kit
871
0.94 ng/mL
Storage is content dependent.
SERPINH1
Biological Fluids, Plasma, Serum