Learn More
Novus Biologicals™ Rat IFI16 ELISA Kit (Colorimetric)
Colorimetric ELISA Kit
Supplier: Novus Biologicals™ NBP306815
This item is not returnable.
View return policy
Specifications
15.63 to 1000 pg/mL | |
ELISA Kit (Colorimetric) | |
This kit recognizes Rat IFI16 in samples. No significant cross-reactivity or interference between Rat IFI16 and analogues was observed. | |
3428 | |
1 Kit | |
Storage of components varies. See protocol for specific instructions. | |
Rat |
9.38 pg/mL | |
Biological Fluids, Cell Culture Supernatant, Plasma, Serum | |
ELISA, Sandwich ELISA | |
IFI16 | |
Cytokine Research | |
ifi-16, IFNGIP1gamma-interferon-inducible protein 16, interferon, gamma-inducible protein 16, interferon-gamma induced protein IFI 16, Interferon-inducible myeloid differentiation transcriptional activator, MGC9466, PYHIN2 |