Learn More
Novus Biologicals Rat IGFBP-2 ELISA Kit (Colorimetric)
This kit recognizes Rat IGFBP-2 in samples.
Manufacturer: Novus Biologicals NBP280240
Description
No significant cross-reactivity or interference between Rat IGFBP-2 and analogues was observed.Specifications
0.156 to 10 ng/mL | |
3485 | |
0.094 ng/mL | |
Rat | |
1 Kit | |
Serum, plasma, and other biological fluids |
ELISA | |
ELISA Kit (Colorimetric) | |
Storage is content dependent | |
IGFBP2 | |
IGFBP-2 |