Learn More
Novus Biologicals Rat IGFBP-6 ELISA Kit (Colorimetric)
This kit recognizes Rat IGFBP6 in samples.
Manufacturer: Novus Biologicals NBP280237
Description
No significant cross-reactivity or interference between Rat IGFBP6 and analogues was observed.Specifications
0.156 to 10 ng/mL | |
3489 | |
0.094 ng/mL | |
Rat | |
IGFBP6 | |
IGFBP-6 |
ELISA | |
ELISA Kit (Colorimetric) | |
Storage is content dependent | |
Cell Cycle and Replication | |
1 Kit | |
Serum, plasma, and other biological fluids |