Learn More
Rat IgG, F(ab)2, Fluorescein, Isotype control, Novus Biologicals™
Rat Isotype Control
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP196800
Catalog No. NBP196800
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
IgG F(ab)2 Isotype Control antibody specifically detects IgG F(ab)2 in Rat samples. It is validated for SDS-Page.
Specifications
Specifications
IgG F(ab)2
LYOPH
SDS-Page 1:100 - 1:2000
Conjugated
IgG
1 mg
Store lyophilized antibody at 4°C in the dark. Aliquot reconstituted liquid and store at -20°C. Avoid freeze-thaw cycles.
Assay by immunoelectrophoresis resulted in a single precipitin arc against anti-Fluorescein, anti-Rat IgG, anti-Rat IgG F(ab)2 and anti-Rat Serum. No reaction was observed against anti-Rat IgG F(c) or anti-Pepsin.
Rat
SDS-Page
Fluorescein
0.01 M Sodium Phosphate, 0.15 M Sodium Chloride, pH 7.2. Stabilizer: 10 mg/mL BSA - Immunoglobulin and Protease free. with 0.01% Thimerosal
Rat
Ion exchange chromatography
RUO
Primary
Reconstitute with 1.0 ml deionized water (or equivalent) to desired concentration.