Learn More
Rat IgG, Fc, Biotin, Isotype control, Novus Biologicals™
Rat Isotype Control
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP197279
Catalog No. NBP197279
Add to cart
Description
Description
IgG Fc antibody specifically detects IgG Fc in Rat samples. It is validated for SDS-Page.
Specifications
Specifications
IgG Fc
LYOPH
SDS-Page 1:100 - 1:2000
Conjugated
IgG
1 mg
Store lyophilized antibody at 4°C in the dark. Aliquot reconstituted liquid and store at -20°C. Avoid freeze-thaw cycles.
Assay by immunoelectrophoresis resulted in a single precipitin arc against anti-biotin, anti-Rat IgG, anti- Rat IgG F(c) and anti-Rat Serum. No reaction was observed against anti-Rat IgG F(ab)2 or anti-Papain.
Rat
SDS-Page
Biotin
Lyophilized from 0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2, 10 mg/mL Bovine Serum Albumin (BSA) - Immunoglobulin and Protease free with 0.01% Sodium Azide
Rat
Ion exchange chromatography
RUO
Primary
Reconstitute with 1.0 ml deionized water (or equivalent) to desired concentration.