Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rat IgG, H/L Chains, Agarose Immobilized, Isotype control, Novus Biologicals™

Goat Polyclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP197265

Catalog No. NBP197265


Add to cart

Description

Description

IgG (H+L) Polyclonal antibody specifically detects IgG (H+L) in Rat samples. It is validated for Immunoprecipitation.
Specifications

Specifications

IgGXXXXX H/L Chains
0.02M Potassium Phosphate, 0.15M Sodium Chloride; pH 7.2
Rat
20mg
Store at 4°C in the dark.
Agarose
Conjugated
H/L Chains
RUO
Primary
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit