Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rat IgG1 Chimeric Isotype Control (MOPC-21), Invitrogen™

Rat Recombinant Monoclonal Antibody

Supplier:  Thermo Scientific MA547826

Encompass

Catalog No. PIMA547826


Add to Cart

Description

Description

Specificity: An antibody of unknown specificity that maybe used as a negative control.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Rat IgG1 Chimeric
Recombinant Monoclonal
1 mg/mL
PBS with 0.02% ProClin 300
Rat
Affinity chromatography
RUO
Liquid
Control
MOPC-21
Unconjugated
Immunoglobulin G; Immunoglobulin G1; ImmunoglobulinG; ImmunoglobulinG1
Proprietary sequence.
200 μg
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
IgG1
Product Suggestions

Product Suggestions

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit