Learn More
Rat IgG1 Isotype Control, Clone: 43414R, R&D Systems™
Rat Monoclonal Antibody
Supplier:  R&D Systems MAB005R
Catalog No. MAB005R
Add to Cart
This item is not returnable. View return policy
Description
Description
IgG1 Monoclonal specifically detects IgG1 in Rat samples. It is validated for Control.
Specifications
Specifications
IgG1
43414R
Control
IgG1, Igh-4, RGD1359539, VH7183
KLH
500 μg
Isotype Control (Primary)
Rat
Purified
Monoclonal
Unconjugated
Supplied as a solution in PBS.
Rat
Protein A or G purified from cell culture supernatant
RUO
Selected for use as rat IgG 1 isotype control. In direct ELISAs, no cross-react with recombinant human (rh) 6Ckine, recombinant mouse (rm) 6Ckine, rhACT II (rhMIP-1 beta ), rhBLC/BCA-1, rmC10, recombinant rat (rr) CINC-1, rrCINC-2 alpha , rrCINC-2 beta , rmCRG-2, rhENA-78, rhEotaxin, rmEotaxin, rhEotaxin 2, rhFractalkine, rmFractalkine, rhGCP-2, rmGCP-2, rhGRO alpha , rhGRO beta , rhGRO gamma , rhHCC-1, rhHCC-4, rhI-309, rhIL-8, rhIL-16, rhIP-10, rmJE, rmKC, rmLymphotactin, rmMARC, rhMCP-1, rhMCP-1 R, rhMCP-2, rhMCP-3, rhMCP-4, rmMCP-5, rhMDC, rmMDC, rhMIG, rmMIG, rhMIP-1 alpha , rmMIP-1 alpha , rmMIP-1 beta , rmMIP-1 gamma , rhMIP-1 delta , rmMIP-2, rhMIP-3 alpha , rrMIP-3 alpha , rhMIP-3 beta , rmMIP-3 beta , recombinant viral (rv) MIP-I, rvMIP-II, rvMIP-III, rhMPIF-1, rhNAP-2, rhParc, rhRANTES, rmRANTES, rhSDF-1 alpha , rmSDF-1 alpha , rhSDF-1 beta , rhTarc, rmTCA-3, rhTeck, or rmTeck.
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
IgG1
Product Suggestions
Product Suggestions