Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Rat IgG1 kappa Isotype Control (eBRG1), Brilliant Ultra Violet™ 805, eBioscience™, Invitrogen™
Rat Isotype Control
Supplier: Invitrogen 368430181

Description
Rat IgG1 is a subclass of immunoglobulin G (IgG) antibodies that are commonly used in research and diagnostics. These antibodies are produced by rats in response to an antigen and can be purified from serum or hybridoma cell culture supernatants. Rat IgG1 is often used as a negative control in experiments, as it does not bind to many mammalian antigens. It is also used in immunohistochemistry, flow cytometry, and western blotting to detect specific antigens. Gene targeting is a powerful tool used to manipulate the rat genome and generate animal models for studying human diseases. With gene targeting, specific genes can be knocked out, inserted, or modified to investigate their role in disease pathogenesis.
Specifications
Rat IgG1 kappa | |
eBRG1 | |
Brilliant Ultraviolet 805 | |
Immunoglobulin G, Immunoglobulin G1, ImmunoglobulinG, ImmunoglobulinG1 | |
Affinity chromatography | |
RUO | |
4°, C, store in dark, DO NOT FREEZE! | |
IgG1 κ |
Control, Flow Cytometry | |
0.2 mg/mL | |
PBS with BSA and 0.09% sodium azide, pH 7.2 | |
Rat | |
50 μg | |
Not Applicable | |
Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction